2-Bromo-2,2-dfluoroacetyl chloride ≥97%

Supplier: Matrix Scientific

006460-5G 006460-25G
101779-098EA 115.38 USD
101779-098 101779-100
2-Bromo-2,2-dfluoroacetyl chloride ≥97%
2-Bromo-2,2-dfluoroacetyl chloride

Hazard: Corrosive/Stored under Argon/Air Sensitive/Refrigerate

Formula: C₂BrClF₂O
Boiling Pt: 50°C
MDL Number: MFCD00069081
CAS Number: 3832-48-2
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 50°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR