Cesium fluoride ≥99%

Supplier: Matrix Scientific

006442-100G 006442-500G
101779-080EA 75.38 USD
101779-080 101779-082
Cesium fluoride ≥99%
Cesium fluoride

TCA listed: T
Hazard: Toxic/Corrosive/Hygroscopic

Formula: CsF
MW: 151.9 g/mol
Boiling Pt: 1251 °C
Melting Pt: 682 °C
MDL Number: MFCD00010960
CAS Number: 13400-13-0
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR