Nitrosium hexafluorophosphate

Supplier: Matrix Scientific

006437-25G 006437-5G
101779-074EA 538.46 USD
101779-074 101779-072
Nitrosium hexafluorophosphate
Nitrosium hexafluorophosphate

TCA listed: T
Hazard: Corrosive/Stored under Argon/Hygroscopic/Refrigerate

Formula: F₆NOP
MDL Number: MFCD00040324
CAS Number: 16921-91-8
UN: 3260
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR