N,O-Bis(trimethylsilyl)trifluoroacetamide ≥98%

Supplier: Matrix Scientific

Synonyms: Trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)acetimidate, Bis(Trimethylsilyl)Trifluoroacetamide, BSTFA

006393-25G 006393-100G
101779-038EA 29.23 USD
101779-038 101779-040
N,O-Bis(trimethylsilyl)trifluoroacetamide ≥98%
N,O-Bis(trimethylsilyl)trifluoroacetamide

Hazard: Flammable/Corrosive/Refrigerate

Formula: C₈H₁₈F₃NOSi₂
MW: 257.4 g/mol
Boiling Pt: 45…50 °C (15 mmHg)
Density: 0.971
MDL Number: MFCD00008269
CAS Number: 25561-30-2
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Boiling point 45-50°/15m
Density 0.971
ND20 1.383

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR