Bis(2-methoxyethyl)aminosulfur trifluoride ≥95%

Supplier: Matrix Scientific

006320-5G 006320-25G 006320-100G
101779-016EA 69.23 USD
101779-016 101779-018 101779-020
Bis(2-methoxyethyl)aminosulfur trifluoride ≥95%
Bis(2-methoxyethyl)aminosulfur trifluoride

Stable alternative to DAST


Purity: 95%
CAS: 202289-38-1
MDL Number: MFCD01321415
Molecular Formula: C6H14F3NO2S
Molecular Weight: 221,24
Boiling Point: >80°C(dec)
Hazard: Corrosive/Water Reactive/Refrigerate

Formula: C₆H₁₄F₃NO₂S
MW: 221.24 g/mol
Boiling Pt: >80 °C (dec.)
Density: 1.2
MDL Number: MFCD01321415
CAS Number: 202289-38-1

Order Now

Specification Test Results

Boiling point >80°(dec)
Density 1.2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR