2-Iodoperfluoropropane ≥97%
Supplier: Matrix Scientific
006272-100G
006272-5G
101778-994EA
173.85
USD
101778-994
101778-990
2-Iodoperfluoropropane ≥97%
2-Iodoperfluoropropane
TCA listed: T
Hazard: Toxic/Light Sensitive/Refrigerate
Formula:
C₃F₇I Boiling Pt: 38 °C Melting Pt: –56 °C Density: 2.1 |
MDL Number:
MFCD00042089 CAS Number: 677-69-0 |
Specification Test Results
Boiling point | 38° |
Density | 2.1 |
ND20 | 1.3298 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...