1H,1H-Heptafluoro-n-butyl methacrylate ≥97%
Supplier: Matrix Scientific
Synonyms:
2,2,3,3,4,4,4-Heptafluorobutyl Methacrylate, 1H,1h-heptafluorobutylmethacrylate, Methacrylic acid 2,2,3,3,4,4,4-Heptafluorobutyl ester
006255-5G
006255-25G
101778-978EA
89.23
USD
101778-978
101778-980
1H,1H-Heptafluoro-n-butyl methacrylate ≥97%
1H,1H-Heptafluoro-n-butyl methacrylate
TCA listed: T
Hazard: Flammable/Irritant/Refrigerate/Lachrymator
Formula:
C₈H₇F₇O₂ MW: 268.13 g/mol Boiling Pt: 134…136 °C |
MDL Number:
MFCD00039251 CAS Number: 13695-31-3 |
Specification Test Results
Boiling point | 134-136° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...