2-(Fluorosulfonyl)difluoroacetyl fluoride ≥97%

Supplier: Matrix Scientific

006229-100G 006229-25G
101778-970EA 1915.38 USD
101778-970 101778-968
2-(Fluorosulfonyl)difluoroacetyl fluoride ≥97%
2-(Fluorosulfonyl)difluoroacetyl fluoride

TCA listed: T
Hazard: Toxic by inhalation/Corrosive

Formula: C₂F₄O₃S
MW: 180.08 g/mol
Boiling Pt: 30 °C
Melting Pt: –110 °C
Density: 1.58
MDL Number: MFCD02093337
CAS Number: 677-67-8

Order Now

Specification Test Results

Boiling point 30°
Density 1.58


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR