2,2-Difluoro-2-(fluorosulfonyl)acetic acid

Supplier: Matrix Scientific

006228-5G 006228-25G
101778-964EA 29.23 USD
101778-964 101778-966
2,2-Difluoro-2-(fluorosulfonyl)acetic acid
2,2-Difluoro-2-(fluorosulphonyl)acetic acid

Hazard: Corrosive/Refrigerate

Formula: C₂HF₃O₄S
Boiling Pt: 88-90°C/45mm
Density: 1.72
MDL Number: MFCD01320807
CAS Number: 1717-59-5

Order Now

Specification Test Results

Boiling point 88-90°/45m

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR