Tetrafluorosuccinic anhydride ≥97%
Supplier: Matrix Scientific
006155-5G
006155-25G
101778-918EA
146.15
USD
101778-918
101778-920
Tetrafluorosuccinic anhydride ≥97%
Tetrafluorosuccinic anhydride
TCA listed: T
Hazard: Toxic by inhalation/Corrosive/Irritant/Moisture Sensitive
Formula:
C₄F₄O₃ MW: 172.04 g/mol Boiling Pt: 54…55 °C Density: 1.6209 |
MDL Number:
MFCD00037917 CAS Number: 699-30-9 |
Specification Test Results
Boiling point | 54-55° |
Density | 1.6209 |
ND20 | 1.324 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...