Pentafluoropyridine ≥99%

Supplier: Matrix Scientific

Synonyms: 2,3,4,5,6-Perfluoropyridine, 2,3,4,5,6-Pentafluoropyridine

006151-100G 006151-25G
101778-912EA 460 USD
101778-912 101778-910
Pentafluoropyridine ≥99%
Pentafluoropyridine

Hazard: Flammable/Irritant

Formula: C₅F₅N
MW: 169.05 g/mol
Boiling Pt: 83-85°C
Density: 1.54
MDL Number: MFCD00006225
CAS Number: 700-16-3
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 83-85°
Density 1.54
ND20 1.386

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR