3-Chloropropyl isothiocyanate ≥98%

Supplier: Matrix Scientific

006140-1G 006140-5G
101778-904EA 33.85 USD
101778-904 101778-906
3-Chloropropyl isothiocyanate ≥98%
3-Chloropropyl isothiocyanate

Hazard: Toxic/Corrosive

Formula: C₄H₆ClNS
MW: 135.62 g/mol
Boiling Pt: 130°C/35mm
Density: 1.18
MDL Number: MFCD00041131
CAS Number: 2799-72-6

Order Now

Specification Test Results

Boiling point 130°/35mm
Density 1.18

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR