(Pentafluorophenyl)acetonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 2,3,4,5,6-Pentafluorobenzyl cyanide, 2-(Perfluorophenyl)acetonitrile, Pentafluorobenzyl cyanide, (2,3,4,5,6-Pentafluorophenyl)acetonitrile

006117-1G 006117-5G 006117-25G
101778-874EA 21.54 USD
101778-874 101778-876 101778-878
(Pentafluorophenyl)acetonitrile ≥97%
(Pentafluorophenyl)acetonitrile

Hazard: Toxic/Irritant

Formula: (C₆F₅)CH₂CN
Boiling Pt: 107…111 °C (17 mmHg)
Density: 1.61
MDL Number: MFCD00013814
CAS Number: 653-30-5
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 107-111°/1
Density 1.61
ND20 1.439


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR