3-(4-Fluorophenyl)propionic acid ≥97%

Supplier: Matrix Scientific

006104-5G 006104-25G
101778-872EA 69.23 USD
101778-872 102832-776
3-(4-Fluorophenyl)propionic acid ≥97%
3-(4-Fluorophenyl)propionic acid

Hazard: Irritant

Formula: C₉H₉FO₂
Melting Pt: 87…88 °C
MDL Number: MFCD00060327
CAS Number: 459-31-4

Order Now

Specification Test Results

Melting point 87-88°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR