1-(3-Fluorophenyl)biguanide hydrochloride ≥97%
Supplier: Matrix Scientific
006085-5G
006085-1G
101778-862EA
200
USD
101778-862
101778-860
1-(3-Fluorophenyl)biguanide hydrochloride ≥97%
1-(3-Fluorophenyl)biguanide hydrochloride
Hazard: Irritant
Formula:
C₈H₁₁ClFN₅ |
MDL Number:
MFCD00203529 CAS Number: 2267-49-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...