2-(4-Fluorophenoxy)-3-nitropyridine ≥97%

Supplier: Matrix Scientific

006062-1G 006062-5G
101778-856EA 176.92 USD
101778-856 101778-858
2-(4-Fluorophenoxy)-3-nitropyridine ≥97%
2-(4-Fluorophenoxy)-3-nitropyridine

Hazard: Toxic

Formula: C₁₁H₇FN₂O₃
Melting Pt: 98-99°C
MDL Number: MFCD00067801
CAS Number: 147143-58-6

Order Now

Specification Test Results

Melting point 98-99°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR