2-(2-Fluorophenoxy)-3-nitropyridine ≥97%
Supplier: Matrix Scientific
006061-1G
006061-5G
101778-852EA
70.77
USD
101778-852
101778-854
2-(2-Fluorophenoxy)-3-nitropyridine ≥97%
2-(2-Fluorophenoxy)-3-nitropyridine
Hazard: Toxic
Formula:
C₁₁H₇FN₂O₃ Melting Pt: 90-92°C |
MDL Number:
MFCD00067805 CAS Number: 175135-65-6 |
Specification Test Results
Melting point | 90-92° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...