Pentafluorophenol ≥99%

Supplier: Matrix Scientific

Synonyms: 2,3,4,5,6-Pentafluorophenol

006058-25G 006058-100G
101778-846EA 96.92 USD
101778-846 101778-848
Pentafluorophenol ≥99%
Pentafluorophenol

TCA listed: T
Hazard: Toxic/Corrosive

Formula: C₆HF₅O
MW: 184.07 g/mol
Boiling Pt: 143 °C
Melting Pt: 34…36 °C
Density: 1.757
MDL Number: MFCD00002156
CAS Number: 771-61-9
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Boiling point 143°
Density 1.757

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR