2-Trifluoromethyl-2-propanol ≥95%

Supplier: Matrix Scientific

006027-5G 006027-25G
101778-830EA 110.77 USD
101778-830 101778-832
2-Trifluoromethyl-2-propanol ≥95%
2-Trifluoromethyl-2-propanol

Hazard: Flammable

Formula: C₄H₇F₃O
Boiling Pt: 80…81 °C
MDL Number: MFCD00014391
CAS Number: 507-52-8

Order Now

Specification Test Results

Boiling point 80-81°
ND20 1.3349


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR