2-(Difluoromethoxy)aniline ≥98%

Supplier: Matrix Scientific

006015-25G 006015-5G
101778-824EA 204.62 USD
101778-824 101778-822
2-(Difluoromethoxy)aniline ≥98%
2-(Difluoromethoxy)aniline

Hazard: Toxic

Formula: C₇H₇F₂NO
MW: 159.14 g/mol
Boiling Pt: 205 °C
Density: 1.272
MDL Number: MFCD00190104
CAS Number: 22236-04-0
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 205°
Density 1.272
ND20 1.505

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR