Pentafluoroiodoethane ≥97%

Supplier: Matrix Scientific

005980-25G 005980-100G
101778-790EA 192.31 USD
101778-790 101778-792
Pentafluoroiodoethane ≥97%
Pentafluoroiodoethane

TCA listed: T
Hazard: Irritant/Nonflammable Gas/Store in Freezer

Formula: C₂F₅I
Boiling Pt: 12…13 °C
Density: 2.085
Flash Pt: none
MDL Number: MFCD00039400
CAS Number: 354-64-3
UN: 1956
ADR: 2,

Order Now

Specification Test Results

Boiling point 12-13°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR