Heptafluorobutyric acid ≥98%

Supplier: Matrix Scientific

005972-25G 005972-100G
101778-776EA 44.62 USD
101778-776 101778-778
Heptafluorobutyric acid ≥98%
Heptafluorobutyric acid

TCA listed: T
Hazard: Corrosive/Stench

Formula: C₄HF₇O₂
Boiling Pt: 120 °C (755 mmHg)
Density: 1.645
MDL Number: MFCD00004171
CAS Number: 375-22-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 120°/755mm
Density 1.645


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR