3,3,4,4,4-Pentafluoro-2-butanol ≥95%
Supplier: Matrix Scientific
005967-1G
005967-5G
101778-768EA
52.31
USD
101778-768
101778-770
3,3,4,4,4-Pentafluoro-2-butanol ≥95%
3,3,4,4,4-Pentafluoro-2-butanol
Hazard: Flammable
Formula:
C₄H₅F₅O MW: 164.08 g/mol Boiling Pt: 83…85 °C |
MDL Number:
MFCD00042432 CAS Number: 374-40-3 |
Specification Test Results
Boiling point | 83-85° |
ND20 | 1.315 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...