2-(2-Chloro-6-fluorobenzylthio)ethylamine ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Aminoethyl 2-chloro-6-fluorobenzyl sulphide

005932-5G 005932-1G
101778-752EA 267.69 USD
101778-752 101778-750
2-(2-Chloro-6-fluorobenzylthio)ethylamine ≥97%
2-(2-Chloro-6-fluorobenzylthio)ethylamine

Hazard: Corrosive

Formula: C₉H₁₁ClFNS
MW: 219.71 g/mol
Boiling Pt: 140°C/6mm
MDL Number: MFCD00052683
CAS Number: 175136-76-2

Order Now

Specification Test Results

Boiling point 140°/6mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR