4,4,5,5,6,6,6-Heptafluoro-3,3-bis(trifluoromethyl)-1-hexene ≥97%
Supplier: Matrix Scientific
005906-5G
005906-25G
101778-740EA
203.08
USD
101778-740
101778-742
4,4,5,5,6,6,6-Heptafluoro-3,3-bis(trifluoromethyl)-1-hexene ≥97%
4,4,5,5,6,6,6-Heptafluoro-3,3-bis(trifluoromethyl)-1-hexene
Hazard: Irritant
Formula:
C₈H₃F₁₃ Boiling Pt: 101…102 °C |
MDL Number:
MFCD00155806 CAS Number: 79272-26-7 |
Specification Test Results
Boiling point | 101-102° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...