α,α,α,α',α',α'-Hexafluoro-2,5-xylidine ≥97%

Supplier: Matrix Scientific

005879-100G 005879-25G
101778-724EA 75.38 USD
101778-724 101778-722
α,α,α,α',α',α'-Hexafluoro-2,5-xylidine ≥97%
α,α,α,α',α',α'-Hexafluoro-2,5-xylidine

Hazard: Toxic

Formula: C₈H₅F₆N
Boiling Pt: 70…71 °C (15 mmHg)
Density: 1.467
MDL Number: MFCD00074940
CAS Number: 328-93-8
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 70-71°/15m
Density 1.467
ND20 1.432


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR