3,5-Bis(trifluoromethyl)thiophenol ≥95%
Supplier: Matrix Scientific
Synonyms:
3,5-Bis(trifluoromethyl)benzenethiol
005876-5G
005876-25G
101778-714EA
115.38
USD
101778-714
101778-716
3,5-Bis(trifluoromethyl)thiophenol ≥95%
3,5-Bis(trifluoromethyl)thiophenol
Hazard: Toxic/Stench/Air Sensitive
Formula:
C₈H₄F₆S MW: 246.18 g/mol Boiling Pt: 64 °C (15 mmHg) |
MDL Number:
MFCD00042273 CAS Number: 130783-02-7 UN: 2810 ADR: 6.1,III |
Specification Test Results
Boiling point | 64°/15mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...