3,5-Bis(trifluoromethyl)benzonitrile ≥97%

Supplier: Matrix Scientific

005871-25G 005871-100G
101778-710EA 72.31 USD
101778-710 101778-712
3,5-Bis(trifluoromethyl)benzonitrile ≥97%
3,5-Bis(trifluoromethyl)benzonitrile

Hazard: Toxic/Irritant/Refrigerate

Formula: C₉H₃F₆N
MW: 239.12 g/mol
Boiling Pt: 155 °C
Density: 1.42
MDL Number: MFCD00000379
CAS Number: 27126-93-8
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 155°
Density 1.42
ND20 1.4175

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR