3,3,3-Trifluoro-2-hydroxy-2-(trifluoromethyl)propionic acid ≥95%

Supplier: Matrix Scientific

005869-5G 005869-1G 005869-25G
101778-704EA 386.15 USD
101778-704 101778-702 101778-706
3,3,3-Trifluoro-2-hydroxy-2-(trifluoromethyl)propionic acid ≥95%
3,3,3-Trifluoro-2-hydroxy-2-(trifluoromethyl)propionic acid

Hazard: Corrosive/Irritant

Formula: C₄H₂F₆O₃
Boiling Pt: 153…156 °C
Melting Pt: 30 °C
MDL Number: MFCD00190631
CAS Number: 662-22-6

Order Now

Specification Test Results

Boiling point 153-156°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR