α,α,α,α',α',α'-Hexafluoro-m-xylene ≥99%

Supplier: Matrix Scientific

Synonyms: 1,3-Bis(trifluoromethyl)benzene, α,α,α,β,β,β-Hexafluoro-m-xylene

005866-100G 005866-500G
101778-694EA 61.54 USD
101778-694 101778-696
α,α,α,α',α',α'-Hexafluoro-m-xylene ≥99%
α,α,α,α',α',α'-Hexafluoro-m-xylene

TCA listed: T
Hazard: Flammable

Formula: C₈H₄F₆
MW: 214.11 g/mol
Boiling Pt: 116…116.3 °C
Density: 1.378
MDL Number: MFCD00000392
CAS Number: 402-31-3
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 116-116.3°
Density 1.378
ND20 1.379

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR