3,5-Bis(trifluoromethyl)benzophenone ≥97%

Supplier: Matrix Scientific

005865-1G 005865-5G
101778-690EA 26.15 USD
101778-690 101778-692
3,5-Bis(trifluoromethyl)benzophenone ≥97%
3,5-Bis(trifluoromethyl)benzophenone

Hazard: Irritant

Formula: C₁₅H₈F₆O
Boiling Pt: 112°C/0.2mm
MDL Number: MFCD00015510
CAS Number: 21221-93-2

Order Now

Specification Test Results

Boiling point 112°/0.2mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR