3,5-Bis(trifluoromethyl)phenyl isothiocyanate ≥99%

Supplier: Matrix Scientific

005864-5G 005864-25G
101778-686EA 69.23 USD
101778-686 101778-688
3,5-Bis(trifluoromethyl)phenyl isothiocyanate ≥99%
3,5-Bis(trifluoromethyl)phenyl isothiocyanate

Hazard: Toxic/Corrosive/Refrigerate

Formula: C₉H₃F₆NS
Boiling Pt: 110°C/35mm
Density: 1.48
MDL Number: MFCD00040838
CAS Number: 23165-29-9

Order Now

Specification Test Results

Boiling point 110°/35mm
Density 1.48


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR