α,α,α,α',α',α'-Hexafluoro-2,4-xylidine

Supplier: Matrix Scientific

Synonyms: 2,4-Bis(trifluoromethyl)aniline, 2,4-Ditrifluoromethylaniline

005838-1G 005838-5G
101778-680EA 84.62 USD
101778-680 101778-682
α,α,α,α',α',α'-Hexafluoro-2,4-xylidine
α,α,α,α',α',α'-Hexafluoro-2,4-xylidine

Hazard: Toxic

Formula: C₈H₅F₆N
MW: 229.12
Boiling Pt: 50 °C (0.4 Torr)
MDL Number: MFCD00236209
CAS Number: 367-71-5
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR