1,4-Bis(trifluoromethyl)-2-nitrobenzene ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Nitro-1,4-bis(trifluoromethyl)benzene, 1-Nitro-2,5-bis(trifluoromethyl)benzene

005832-5G 005832-25G
101778-676EA 21.54 USD
101778-676 101778-678
1,4-Bis(trifluoromethyl)-2-nitrobenzene ≥97%
1,4-Bis(trifluoromethyl)-2-nitrobenzene

Hazard: Toxic

Formula: C₈H₃F₆NO₂
MW: 259.11 g/mol
Boiling Pt: 83 °C (11 Torr)
Density: 1.57
Flash Pt: 76 °C
MDL Number: MFCD00236674
CAS Number: 320-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR