3,5-Bis(trifluoromethyl)benzyl chloride ≥97%

Supplier: Matrix Scientific

005831-100G 005831-25G
101778-674EA 330.77 USD
101778-674 101778-672
3,5-Bis(trifluoromethyl)benzyl chloride ≥97%
3,5-Bis(trifluoromethyl)benzyl chloride

Hazard: Corrosive

Formula: C₉H₅ClF₆
Melting Pt: 30-32°C
MDL Number: MFCD00009906
CAS Number: 75462-59-8
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Melting point 30-32°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR