3,5-Bis(trifluoromethyl)benzylamine ≥97%

Supplier: Matrix Scientific

Synonyms: (3,5-Bis(trifluoromethyl)phenyl)methanamine

005828-100G 005828-25G
101778-666EA 653.85 USD
101778-666 101778-664
3,5-Bis(trifluoromethyl)benzylamine ≥97%
3,5-Bis(trifluoromethyl)benzylamine

Hazard: Corrosive

Formula: C₉H₇F₆N
MW: 243.15 g/mol
Melting Pt: 50…55 °C
MDL Number: MFCD00009909
CAS Number: 85068-29-7

Order Now

Specification Test Results

Melting point 50-55°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR