3,3-Bis(trifluoromethyl)-3-hydroxypropionic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Hydroxy-2H,2H-perfluoro-3-methylbutanoic acid
005812-1G
005812-5G
101778-640EA
44.62
USD
101778-640
101778-642
3,3-Bis(trifluoromethyl)-3-hydroxypropionic acid ≥97%
3,3-Bis(trifluoromethyl)-3-hydroxypropionic acid
Hazard: Corrosive
Formula:
C₅H₄F₆O₃ MW: 226.08 g/mol Melting Pt: 85…89 °C |
MDL Number:
MFCD00040910 CAS Number: 1547-36-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...