3,5-Bis(trifluoromethyl)benzyl bromide ≥97%
Supplier: Matrix Scientific
Synonyms:
1-(Bromomethyl)-3,5-bis(trifluoromethyl)benzene
005809-5G
005809-25G
101778-634EA
36.92
USD
101778-634
101778-636
3,5-Bis(trifluoromethyl)benzyl bromide ≥97%
3,5-Bis(trifluoromethyl)benzyl bromide
Hazard: Flammable/Corrosive/Refrigerate/Lachrymator
Formula:
C₉H₅BrF₆ MW: 307.03 g/mol Boiling Pt: 45…47 °C (10 mmHg) Density: 1.675 Flash Pt: 26 °C |
MDL Number:
MFCD00009905 CAS Number: 32247-96-4 UN: 3265 ADR: 8,II |
Specification Test Results
Density | 1.675 |
ND20 | 1.445 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...