7-Bromoheptanenitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 7-Bromoheptanonitrile

005754-5G 005754-25G
101778-608EA 184.62 USD
101778-608 101778-610
7-Bromoheptanenitrile ≥95%
7-Bromoheptanenitrile

TCA listed: T
Hazard: Toxic/Irritant

Formula: BrH₂C(CH₂)₄CH₂CN
MW: 190.08 g/mol
Boiling Pt: 140-141°C/14mm
Density: 1.265
MDL Number: MFCD00001982
CAS Number: 20965-27-9
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 140-141°/1
Density 1.265
ND20 1.475

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR