3,3,4,4,5,5,5-Heptafluoro-2-pentanone ≥97%
Supplier: Matrix Scientific
005753-25G
005753-5G
101778-606EA
441.54
USD
101778-606
101778-604
3,3,4,4,5,5,5-Heptafluoro-2-pentanone ≥97%
3,3,4,4,5,5,5-Heptafluoro-2-pentanone
Hazard: Flammable
Formula:
C₅H₃F₇O Boiling Pt: 63…64 °C Density: 1.57 |
MDL Number:
MFCD00042079 CAS Number: 355-17-9 |
Specification Test Results
Boiling point | 63-64° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...