2-Iodocumene ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Iodoisopropylbenzene, 1-Iodo-2-isopropylbenzene, 2-(2-Iodophenyl) propane

005746-25G 005746-5G
101778-602EA 186.15 USD
101778-602 101778-600
2-Iodocumene ≥98%
2-Iodocumene

Hazard: Irritant

Formula: C₉H₁₁I
MW: 246.09 g/mol
Boiling Pt: 110°C/20mm
MDL Number: MFCD00013704
CAS Number: 19099-54-8

Order Now

Specification Test Results

Boiling point 110°/20mm
ND20 1.5801

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR