(4-Fluorophenylthio)acetic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
2-((4-Fluorophenyl)thio)acetic acid
005729-5G
005729-25G
101778-596EA
92.31
USD
101778-596
101778-598
(4-Fluorophenylthio)acetic acid ≥97%
(4-Fluorophenylthio)acetic acid
Hazard: Corrosive
Formula:
C₈H₇FO₂S Melting Pt: 76…79 °C |
MDL Number:
MFCD00219825 CAS Number: 332-51-4 |
Specification Test Results
Melting point | 76-79° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...