2-Methoxyphenyl isothiocyanate ≥98%

Supplier: Matrix Scientific

005702-5G 005702-25G
101778-588EA 40 USD
101778-588 101778-590
2-Methoxyphenyl isothiocyanate ≥98%
2-Methoxyphenyl isothiocyanate

Hazard: Toxic/Corrosive/Moisture Sensitive/Lachrymator

Formula: C₈H₇NOS
Boiling Pt: 264…266 °C
Density: 1.196
MDL Number: MFCD00011675
CAS Number: 3288-04-8

Order Now

Specification Test Results

Boiling point 264-266°
Density 1.17
ND20 1.6443


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR