4-(2,3-Epoxypropyl)morpholine ≥98%

Supplier: Matrix Scientific

005672-5G 005672-25G
101778-582EA 80 USD
101778-582 101778-584
4-(2,3-Epoxypropyl)morpholine ≥98%
4-(2,3-Epoxypropyl)morpholine

Hazard: Irritant/Store in Freezer

Formula: C₇H₁₃NO₂
Boiling Pt: 93…98 °C (12 mmHg)
Density: 1.05
MDL Number: MFCD00047415
CAS Number: 6270-19-5

Order Now

Specification Test Results

Boiling point 93-98°/12m
Density 1.05

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR