Benzyl isothiocyanate ≥98%
Supplier: Matrix Scientific
Synonyms:
Benzylisothiocyanate
005639-5G
005639-25G
101778-572EA
36.92
USD
101778-572
101778-574
Benzyl isothiocyanate ≥98%
Benzyl isothiocyanate
TCA listed: T
Hazard: Toxic/Moisture Sensitive/Lachrymator
Formula:
C₈H₇NS Boiling Pt: 242…243 °C Melting Pt: 41 °C Density: 1.125 |
MDL Number:
MFCD00004819 CAS Number: 622-78-6 UN: 2206 ADR: 6.1,II |
Specification Test Results
Boiling point | 242-243° |
Density | 1.125 |
ND20 | 1.6018 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...