2,4-Difluorotoluene ≥99%
Supplier: Matrix Scientific
Synonyms:
2,4-Difluoro-1-methylbenzene
005630-25G
005630-100G
101778-564EA
73.85
USD
101778-564
101778-566
2,4-Difluorotoluene ≥99%
2,4-Difluorotoluene
Hazard: Flammable
Formula:
C₇H₆F₂ MW: 128.12 g/mol Boiling Pt: 114-115°C Density: 1.12 |
MDL Number:
MFCD00000332 CAS Number: 452-76-6 UN: 2388 ADR: 3,II |
Specification Test Results
Boiling point | 114-115° |
Density | 1.12 |
ND20 | 1.449 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...