2,6-Difluorotoluene ≥99%

Supplier: Matrix Scientific

005629-25G 005629-5G
101778-562EA 181.54 USD
101778-562 101778-560
2,6-Difluorotoluene ≥99%
2,6-Difluorotoluene

Hazard: Flammable

Formula: C₇H₆F₂
Boiling Pt: 112 °C (740 mmHg)
Density: 1.129
MDL Number: MFCD00043898
CAS Number: 443-84-5
UN: 2388
ADR: 3,II

Order Now

Specification Test Results

Boiling point 112°/740mm
Density 1.129
ND20 1.453


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR