(3,5-Difluorophenyl)acetonitrile ≥98%
Supplier: Matrix Scientific
005588-5G
005588-25G
101778-526EA
73.85
USD
101778-526
101778-528
(3,5-Difluorophenyl)acetonitrile ≥98%
(3,5-Difluorophenyl)acetonitrile
Hazard: Toxic
Formula:
C₈H₅F₂N Melting Pt: 34…36 °C |
MDL Number:
MFCD00061278 CAS Number: 122376-76-5 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...