(3,4-Difluorophenyl)acetonitrile ≥98%

Supplier: Matrix Scientific

005587-5G 005587-25G
101778-524EA 47.69 USD
101778-524 102550-866
(3,4-Difluorophenyl)acetonitrile ≥98%
(3,4-Difluorophenyl)acetonitrile

Hazard: Toxic

Formula: C₈H₅F₂N
Density: 1.244
MDL Number: MFCD00009974
CAS Number: 658-99-1
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Density 1.244
ND20 1.484


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR