(2,6-Difluorophenyl)acetonitrile ≥98%

Supplier: Matrix Scientific

005586-5G 005586-25G
101778-520EA 36.92 USD
101778-520 101778-522
(2,6-Difluorophenyl)acetonitrile ≥98%
(2,6-Difluorophenyl)acetonitrile

Hazard: Flammable/Toxic

Formula: C₈H₅F₂N
Density: 1.238
MDL Number: MFCD00009973
CAS Number: 654-01-3
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Density 1.238
ND20 1.481


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR